Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616243 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0922 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0922 - KIAA0922 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131007.1 | Intron | NP_001124479.1 | ||||
NM_015196.3 | Intron | NP_056011.3 | ||||
XM_011531780.1 | Intron | XP_011530082.1 | ||||
XM_011531781.1 | Intron | XP_011530083.1 | ||||
XM_011531783.1 | Intron | XP_011530085.1 | ||||
XM_017007924.1 | Intron | XP_016863413.1 | ||||
XM_017007925.1 | Intron | XP_016863414.1 | ||||
XM_017007926.1 | Intron | XP_016863415.1 | ||||
XM_017007927.1 | Intron | XP_016863416.1 |