Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGAGCTGCCCCGCTTGCAAGA[G/T]CCGGAGGAGGACGAGGATTGGTGAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 300369 MIM: 300255 | |||||||||||||||||||||||
Literature Links: |
ACRC PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ACRC - acidic repeat containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052957.4 | 143 | Missense Mutation | GAG,GAT | E,D 13 | NP_443189.1 | |
XM_006724716.3 | 143 | Missense Mutation | GAG,GAT | E,D 13 | XP_006724779.1 | |
XM_011531063.2 | 143 | Intron | XP_011529365.1 | |||
XM_011531064.1 | 143 | Missense Mutation | GAG,GAT | E,D 13 | XP_011529366.1 | |
XM_011531066.2 | 143 | Intron | XP_011529368.1 | |||
XM_011531067.2 | 143 | Intron | XP_011529369.1 | |||
XM_017029965.1 | 143 | Intron | XP_016885454.1 |
OGT - O-linked N-acetylglucosamine (GlcNAc) transferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |