Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACTACATTTTGTTTCTCTATTC[C/T]TTGATTGATTCTTGGGTTATCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603086 MIM: 607607 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ART3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ART3 - ADP-ribosyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP54 - nucleoporin 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278603.1 | Intron | NP_001265532.1 | ||||
NM_017426.3 | Intron | NP_059122.2 | ||||
XM_006714237.1 | Intron | XP_006714300.1 | ||||
XM_011532033.2 | Intron | XP_011530335.1 | ||||
XM_011532034.2 | Intron | XP_011530336.1 | ||||
XM_017008294.1 | Intron | XP_016863783.1 | ||||
XM_017008295.1 | Intron | XP_016863784.1 |
Set Membership: |
HapMap |