Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGTGGTCCTTTCATCTGTTAAT[A/C]CAGTGCTCAACACGTTCAGAGTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 179502 | ||||||||||||||||||||
Literature Links: |
RAP1GDS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAP1GDS1 - Rap1 GTPase-GDP dissociation stimulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100426.1 | Intron | NP_001093896.1 | ||||
NM_001100427.1 | Intron | NP_001093897.1 | ||||
NM_001100428.1 | Intron | NP_001093898.1 | ||||
NM_001100429.1 | Intron | NP_001093899.1 | ||||
NM_001100430.1 | Intron | NP_001093900.1 | ||||
NM_021159.4 | Intron | NP_066982.3 |