Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTCTGGTGTATATGAGCTACTT[C/T]GTCTCCTGTGAGAGTGGTATCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603755 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFYVE9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFYVE9 - zinc finger FYVE-type containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004799.3 | Intron | NP_004790.2 | ||||
NM_007324.3 | Intron | NP_015563.2 | ||||
XM_011542437.2 | Intron | XP_011540739.1 | ||||
XM_017002845.1 | Intron | XP_016858334.1 |