Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAAAAGTAAAAATTGTATGAAG[A/G]AGCAGGAAAATTTTCACCATCAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180610 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RLF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RLF - rearranged L-myc fusion | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012421.3 | Intron | NP_036553.2 | ||||
XM_017002016.1 | Intron | XP_016857505.1 | ||||
XM_017002017.1 | Intron | XP_016857506.1 | ||||
XM_017002018.1 | Intron | XP_016857507.1 |
Set Membership: |
HapMap |