Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTTTTGAGGTGCAAAAGTTTTA[A/G]ACTTTGATGAAGTCCAGTTTATCAT
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 614428 | ||||||||||||||||||||||||||
Literature Links: |
TFAP2E PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
TFAP2E - transcription factor AP-2 epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178548.3 | Intron | NP_848643.2 | ||||
XM_017001139.1 | Intron | XP_016856628.1 |
Set Membership: |
HapMap |