Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGAGGCAAAAAGGGGCTATCAA[A/C]GGTGTAGGAAGTAGATTGAGAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608458 MIM: 614428 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCDN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NCDN - neurochondrin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TFAP2E - transcription factor AP-2 epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178548.3 | 228 | Intron | NP_848643.2 | |||
XM_017001139.1 | 228 | UTR 5 | XP_016856628.1 |
Set Membership: |
HapMap |