Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTGTTGTTGTTTGTTTGTTTA[A/G]AAATAACTATTGATAACTGTTAAGA
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 601241 MIM: 153390 | ||||||||||||||||||||||||||
Literature Links: |
HDAC1 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
HDAC1 - histone deacetylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004964.2 | Intron | NP_004955.2 | ||||
XM_011541309.2 | Intron | XP_011539611.1 |
LCK - LCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |