Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATTTCTTTTACATGTGTTCACA[C/T]AGAATTCCAGAGAGAGATCGGCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604409 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMEB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GMEB1 - glucocorticoid modulatory element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319674.1 | Intron | NP_001306603.1 | ||||
NM_006582.3 | Intron | NP_006573.2 | ||||
NM_024482.2 | Intron | NP_077808.1 | ||||
XM_011540518.2 | Intron | XP_011538820.1 | ||||
XM_011540519.2 | Intron | XP_011538821.1 | ||||
XM_011540521.2 | Intron | XP_011538823.1 | ||||
XM_011540522.2 | Intron | XP_011538824.1 | ||||
XM_017000087.1 | Intron | XP_016855576.1 |