Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACTCCCTGTCACTTCACCACACA[C/T]TCCCTTTCAGGAGCTCATTGCCTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607767 | |||||||||||||||||||||||
Literature Links: |
SESN2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SESN2 - sestrin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031459.4 | Intron | NP_113647.1 |
Set Membership: |
HapMap |