Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGAGGCCACGATAACTGCACTA[G/T]TGGAGCCCTGACTCAGACACCCCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187020 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCP10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCP10 - t-complex 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004610.3 | 1698 | Intron | NP_004601.3 | |||
XM_011536082.2 | 1698 | Intron | XP_011534384.1 | |||
XM_011536084.2 | 1698 | Intron | XP_011534386.1 | |||
XM_011536085.2 | 1698 | Intron | XP_011534387.1 | |||
XM_011536088.2 | 1698 | Intron | XP_011534390.1 | |||
XM_011536090.2 | 1698 | Intron | XP_011534392.1 | |||
XM_017011228.1 | 1698 | Intron | XP_016866717.1 | |||
XM_017011229.1 | 1698 | Intron | XP_016866718.1 | |||
XM_017011230.1 | 1698 | Intron | XP_016866719.1 | |||
XM_017011231.1 | 1698 | UTR 3 | XP_016866720.1 |