Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTGTTGCTGGCAGGAGCGTCGT[A/C]TCAGCGCTCGGTGGCCAGAATGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604473 | ||||||||||||||||||||
Literature Links: |
GRM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRM1 - glutamate metabotropic receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278064.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | NP_001264993.1 | |
NM_001278065.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | NP_001264994.1 | |
NM_001278066.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | NP_001264995.1 | |
NM_001278067.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | NP_001264996.1 | |
XM_011535782.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | XP_011534084.1 | |
XM_017010783.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | XP_016866272.1 | |
XM_017010784.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | XP_016866273.1 | |
XM_017010785.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | XP_016866274.1 | |
XM_017010786.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | XP_016866275.1 | |
XM_017010787.1 | 580 | Missense Mutation | TAT,TCT | Y,S 34 | XP_016866276.1 | |
XM_017010788.1 | 580 | Intron | XP_016866277.1 |