Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605668 MIM: 613770 | ||||||||||||||||||||
Literature Links: |
BACE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BACE2 - beta-site APP-cleaving enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012105.4 | Intron | NP_036237.2 | ||||
NM_138991.2 | Intron | NP_620476.1 | ||||
NM_138992.2 | Intron | NP_620477.1 | ||||
XM_017028314.1 | Intron | XP_016883803.1 |
MIR3197 - microRNA 3197 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLAC4 - placenta specific 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |