Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGCGAGTACTTGTTTTTGGACA[C/T]TATGCATCAATTTCTTCTACTTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108962 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NPR3 - natriuretic peptide receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000908.3 | Intron | NP_000899.1 | ||||
NM_001204375.1 | Intron | NP_001191304.1 | ||||
NM_001204376.1 | Intron | NP_001191305.1 | ||||
XM_005248309.1 | Intron | XP_005248366.1 | ||||
XM_011514047.1 | Intron | XP_011512349.1 | ||||
XM_011514048.2 | Intron | XP_011512350.1 | ||||
XM_011514049.2 | Intron | XP_011512351.1 | ||||
XM_011514050.2 | Intron | XP_011512352.1 | ||||
XM_017009492.1 | Intron | XP_016864981.1 |
Set Membership: |
HapMap |