Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTGCAGTCTCCATGAAAGTGCA[C/T]ATAAACGGTTAAGGCAAAGTACCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604653 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC40A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC40A1 - solute carrier family 40 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014585.5 | 3332 | UTR 3 | NP_055400.1 | |||
XM_005246505.1 | 3332 | UTR 3 | XP_005246562.1 | |||
XM_017003938.1 | 3332 | UTR 3 | XP_016859427.1 |
Set Membership: |
HapMap |