Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACATGTAGTACTCAGAGCTCTA[C/T]GGAAACCCAGGCACCTCGACCTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612131 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DHRS9 - dehydrogenase/reductase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142270.1 | 303 | Intron | NP_001135742.1 | |||
NM_001142271.1 | 303 | Intron | NP_001135743.1 | |||
NM_001289763.1 | 303 | Intron | NP_001276692.1 | |||
NM_199204.1 | 303 | UTR 5 | NP_954674.1 | |||
XM_011510475.1 | 303 | UTR 5 | XP_011508777.1 |
Set Membership: |
HapMap |