Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTATACTTGCTTATTCAGACTC[A/G]TTTTCTATACTTTCCTCAAGATACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600403 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAP - fibroblast activation protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291807.1 | Intron | NP_001278736.1 | ||||
NM_004460.3 | Intron | NP_004451.2 | ||||
XM_011510796.2 | Intron | XP_011509098.1 | ||||
XM_011510797.2 | Intron | XP_011509099.1 | ||||
XM_017003585.1 | Intron | XP_016859074.1 |
LOC101929532 - uncharacterized LOC101929532 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |