Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCAGCCCTGAAGTAATCTGTG[C/T]GAGCAGCTGAGGCAGGCTCTGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300859 MIM: 300292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC22 - coiled-coil domain containing 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014008.4 | 1804 | Intron | NP_054727.1 | |||
XM_005272599.3 | 1804 | Intron | XP_005272656.1 |
FOXP3 - forkhead box P3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114377.1 | 1804 | UTR 3 | NP_001107849.1 | |||
NM_014009.3 | 1804 | UTR 3 | NP_054728.2 | |||
XM_006724533.2 | 1804 | UTR 3 | XP_006724596.2 | |||
XM_011543916.2 | 1804 | UTR 3 | XP_011542218.1 | |||
XM_017029565.1 | 1804 | UTR 3 | XP_016885054.1 | |||
XM_017029566.1 | 1804 | UTR 3 | XP_016885055.1 | |||
XM_017029567.1 | 1804 | UTR 3 | XP_016885056.1 |