Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAGCTCCGCTGAGAGTCTGAC[C/T]TCAGTTGAATTAAATTTGACCTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609098 | ||||||||||||||||||||
Literature Links: |
FBXO25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO25 - F-box protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012173.3 | Intron | NP_036305.2 | ||||
NM_183420.1 | Intron | NP_904356.1 | ||||
NM_183421.1 | Intron | NP_904357.1 | ||||
XM_011534748.2 | Intron | XP_011533050.1 | ||||
XM_011534749.1 | Intron | XP_011533051.1 | ||||
XM_017013313.1 | Intron | XP_016868802.1 | ||||
XM_017013314.1 | Intron | XP_016868803.1 |