Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATTGTGTATTTCTATATTTGCA[A/T]AGAGTACTTTAGAACTTTTTATGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609613 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHM2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLEKHM2 - pleckstrin homology and RUN domain containing M2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015164.2 | Intron | NP_055979.2 | ||||
XM_005245790.3 | Intron | XP_005245847.1 | ||||
XM_005245791.4 | Intron | XP_005245848.1 | ||||
XM_017000757.1 | Intron | XP_016856246.1 | ||||
XM_017000758.1 | Intron | XP_016856247.1 |