Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCAGTTAGGGACTGTTAGAAGA[C/T]GGTCGCATTTTTGGGGGTTGGGTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614392 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TDRD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TDRD3 - tudor domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146070.1 | Intron | NP_001139542.1 | ||||
NM_001146071.1 | Intron | NP_001139543.1 | ||||
NM_030794.2 | Intron | NP_110421.1 | ||||
XM_005266556.4 | Intron | XP_005266613.1 | ||||
XM_005266560.2 | Intron | XP_005266617.1 | ||||
XM_011535247.1 | Intron | XP_011533549.1 | ||||
XM_011535249.2 | Intron | XP_011533551.1 | ||||
XM_017020777.1 | Intron | XP_016876266.1 |
Set Membership: |
HapMap |