Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCCAGCTGCAAAGGCATCTGTA[A/G]GGTGGGATGATTGTCTTAGTCTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126391 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf68 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C19orf68 - chromosome 19 open reading frame 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199341.3 | 346 | Intron | NP_955373.3 | |||
XM_005259449.3 | 346 | Intron | XP_005259506.1 | |||
XM_006723204.3 | 346 | Intron | XP_006723267.1 | |||
XM_006723205.2 | 346 | Intron | XP_006723268.1 | |||
XM_011526936.2 | 346 | Missense Mutation | AGG,GGG | R,G 20 | XP_011525238.1 | |
XM_011526937.2 | 346 | Intron | XP_011525239.1 |
LIG1 - DNA ligase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |