Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAGGCAGGGCTTTTGGGGAAAC[G/T]CTCTGGGGAACATAGACTGGTCCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607315 MIM: 603602 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CABP5 - calcium binding protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G4C - phospholipase A2 group IVC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159322.1 | Intron | NP_001152794.1 | ||||
NM_001159323.1 | Intron | NP_001152795.1 | ||||
NM_003706.2 | Intron | NP_003697.2 | ||||
XM_011527431.2 | Intron | XP_011525733.1 | ||||
XM_011527432.2 | Intron | XP_011525734.1 | ||||
XM_011527433.2 | Intron | XP_011525735.1 | ||||
XM_017027411.1 | Intron | XP_016882900.1 | ||||
XM_017027412.1 | Intron | XP_016882901.1 | ||||
XM_017027413.1 | Intron | XP_016882902.1 | ||||
XM_017027414.1 | Intron | XP_016882903.1 |
Set Membership: |
HapMap |