Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGCCACTAACTAACTTTATTAG[A/G]CTAGTTTTTACATAAATAACCAGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600694 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL6ST PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL6ST - interleukin 6 signal transducer | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190981.1 | 5511 | UTR 3 | NP_001177910.1 | |||
NM_002184.3 | 5511 | UTR 3 | NP_002175.2 | |||
NM_175767.2 | 5511 | UTR 3 | NP_786943.1 | |||
XM_017009441.1 | 5511 | UTR 3 | XP_016864930.1 | |||
XM_017009442.1 | 5511 | Intron | XP_016864931.1 |
Set Membership: |
HapMap |