Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTCCAGGAACAATCTCAGGAA[C/T]CTGCATTTGTAAAAATCCTCCCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615498 MIM: 601243 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIEF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIEF2 - mitochondrial elongation factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOP3A - topoisomerase (DNA) III alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320759.1 | 5013 | UTR 3 | NP_001307688.1 | |||
NM_004618.4 | 5013 | UTR 3 | NP_004609.1 | |||
XM_011524001.2 | 5013 | UTR 3 | XP_011522303.1 |
Set Membership: |
HapMap |