Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAATTTTCAGTGTCCTTAATTTT[A/G]TTATATAAATCATGACAAGTAAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF697 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF697 - zinc finger protein 697 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080470.1 | 6254 | UTR 3 | NP_001073939.1 | |||
XM_005271315.3 | 6254 | UTR 3 | XP_005271372.1 | |||
XM_011542416.2 | 6254 | UTR 3 | XP_011540718.1 | |||
XM_017002783.1 | 6254 | UTR 3 | XP_016858272.1 |