Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AATATATAAATATATAATAAACATA[A/C]CAGACCAGTTAGATATATAATAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF780A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF780A - zinc finger protein 780A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010880.2 | 6465 | Intron | NP_001010880.2 | |||
NM_001142577.1 | 6465 | Intron | NP_001136049.1 | |||
NM_001142578.1 | 6465 | Intron | NP_001136050.1 | |||
NM_001142579.1 | 6465 | UTR 3 | NP_001136051.1 | |||
XM_005258773.2 | 6465 | UTR 3 | XP_005258830.1 | |||
XM_006723150.3 | 6465 | UTR 3 | XP_006723213.1 | |||
XM_011526771.2 | 6465 | UTR 3 | XP_011525073.1 | |||
XM_011526772.2 | 6465 | UTR 3 | XP_011525074.1 | |||
XM_017026611.1 | 6465 | UTR 3 | XP_016882100.1 | |||
XM_017026612.1 | 6465 | UTR 3 | XP_016882101.1 | |||
XM_017026613.1 | 6465 | UTR 3 | XP_016882102.1 | |||
XM_017026614.1 | 6465 | UTR 3 | XP_016882103.1 | |||
XM_017026615.1 | 6465 | UTR 3 | XP_016882104.1 | |||
XM_017026616.1 | 6465 | UTR 3 | XP_016882105.1 | |||
XM_017026617.1 | 6465 | UTR 3 | XP_016882106.1 | |||
XM_017026618.1 | 6465 | UTR 3 | XP_016882107.1 | |||
XM_017026619.1 | 6465 | UTR 3 | XP_016882108.1 |