Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGAGTAGCTAGAGCTGGTTCCT[A/G]TTATTCACAGATTCTGTATTTGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123803 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATF1 - activating transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005171.4 | Intron | NP_005162.1 | ||||
XM_011538386.2 | Intron | XP_011536688.1 | ||||
XM_011538387.2 | Intron | XP_011536689.1 | ||||
XM_011538388.2 | Intron | XP_011536690.1 | ||||
XM_017019331.1 | Intron | XP_016874820.1 | ||||
XM_017019332.1 | Intron | XP_016874821.1 | ||||
XM_017019333.1 | Intron | XP_016874822.1 | ||||
XM_017019334.1 | Intron | XP_016874823.1 | ||||
XM_017019335.1 | Intron | XP_016874824.1 | ||||
XM_017019336.1 | Intron | XP_016874825.1 |