Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTGGGTCCAGTGTGATCTCTTC[C/T]TAGGAAATGGCTTAATATTGTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607025 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MELK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MELK - maternal embryonic leucine zipper kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256685.1 | Intron | NP_001243614.1 | ||||
NM_001256687.1 | Intron | NP_001243616.1 | ||||
NM_001256688.1 | Intron | NP_001243617.1 | ||||
NM_001256689.1 | Intron | NP_001243618.1 | ||||
NM_001256690.1 | Intron | NP_001243619.1 | ||||
NM_001256691.1 | Intron | NP_001243620.1 | ||||
NM_001256692.1 | Intron | NP_001243621.1 | ||||
NM_001256693.1 | Intron | NP_001243622.1 | ||||
NM_014791.3 | Intron | NP_055606.1 | ||||
XM_011518076.2 | Intron | XP_011516378.1 | ||||
XM_011518077.1 | Intron | XP_011516379.1 | ||||
XM_011518078.2 | Intron | XP_011516380.1 | ||||
XM_011518079.1 | Intron | XP_011516381.1 | ||||
XM_011518080.1 | Intron | XP_011516382.1 | ||||
XM_011518081.2 | Intron | XP_011516383.1 | ||||
XM_011518082.2 | Intron | XP_011516384.1 | ||||
XM_011518083.2 | Intron | XP_011516385.1 | ||||
XM_011518084.2 | Intron | XP_011516386.1 | ||||
XM_011518085.1 | Intron | XP_011516387.1 | ||||
XM_011518086.2 | Intron | XP_011516388.1 |
Set Membership: |
HapMap |