Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCCCCCCATGCAAGAGTTATA[C/T]CCCATATTTCTCTTTGGAGATTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100880 MIM: 609631 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACO1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACO1 - aconitase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDX58 - DEXD/H-box helicase 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014314.3 | Intron | NP_055129.2 |
LOC101060445 - uncharacterized LOC101060445 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |