Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCAGAGAGCAGTTTAGCTTAGT[A/T]AAGTTTTACATAAATCCTATTGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608040 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT74 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFT74 - intraflagellar transport 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099222.1 | 2522 | Intron | NP_001092692.1 | |||
NM_001099223.1 | 2522 | Intron | NP_001092693.1 | |||
NM_001099224.1 | 2522 | Intron | NP_001092694.1 | |||
NM_025103.2 | 2522 | Intron | NP_079379.2 | |||
XM_011518035.2 | 2522 | Intron | XP_011516337.1 | |||
XM_011518036.2 | 2522 | Intron | XP_011516338.1 | |||
XM_017015163.1 | 2522 | Intron | XP_016870652.1 | |||
XM_017015164.1 | 2522 | Intron | XP_016870653.1 |
LRRC19 - leucine rich repeat containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022901.2 | 2522 | UTR 3 | NP_075052.1 |