Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605469 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM4C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KDM4C - lysine demethylase 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146695.1 | Intron | NP_001140167.1 | ||||
NM_001146696.1 | Intron | NP_001140168.1 | ||||
NM_001304339.1 | Intron | NP_001291268.1 | ||||
NM_001304340.1 | Intron | NP_001291269.1 | ||||
NM_001304341.1 | Intron | NP_001291270.1 | ||||
NM_015061.3 | Intron | NP_055876.2 | ||||
XM_006716741.2 | Intron | XP_006716804.1 | ||||
XM_011517811.2 | Intron | XP_011516113.1 | ||||
XM_011517812.2 | Intron | XP_011516114.2 | ||||
XM_011517816.2 | Intron | XP_011516118.1 | ||||
XM_017014498.1 | Intron | XP_016869987.1 | ||||
XM_017014499.1 | Intron | XP_016869988.1 | ||||
XM_017014500.1 | Intron | XP_016869989.1 | ||||
XM_017014501.1 | Intron | XP_016869990.1 | ||||
XM_017014502.1 | Intron | XP_016869991.1 | ||||
XM_017014503.1 | Intron | XP_016869992.1 | ||||
XM_017014504.1 | Intron | XP_016869993.1 | ||||
XM_017014505.1 | Intron | XP_016869994.1 | ||||
XM_017014506.1 | Intron | XP_016869995.1 |
Set Membership: |
HapMap |