Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCCCTTTCCTTCTCTCTTTAAA[A/G]TAATGTTATAGCACCTTTGATTTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615211 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UHRF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UHRF2 - ubiquitin like with PHD and ring finger domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152896.2 | Intron | NP_690856.1 | ||||
XM_011517704.2 | Intron | XP_011516006.1 | ||||
XM_011517705.2 | Intron | XP_011516007.1 | ||||
XM_017014253.1 | Intron | XP_016869742.1 | ||||
XM_017014254.1 | Intron | XP_016869743.1 |
Set Membership: |
HapMap |