Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTTCTACCAAGAATTGGACCTG[G/T]ACTCTCTTAACGAAGATCTTCTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TPD52L3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPD52L3 - tumor protein D52-like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001874.2 | 215 | UTR 5 | NP_001001874.1 | |||
NM_001001875.3 | 215 | UTR 5 | NP_001001875.1 | |||
NM_033516.5 | 215 | UTR 5 | NP_277051.3 | |||
XM_017015280.1 | 215 | UTR 5 | XP_016870769.1 |