Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCGCTTAATCAGAAGTTTCGTC[A/G]GGCAGTTTCAGAGTTGGTGTCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608678 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL33 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL33 - interleukin 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199640.1 | Intron | NP_001186569.1 | ||||
NM_001199641.1 | Intron | NP_001186570.1 | ||||
NM_001314044.1 | Intron | NP_001300973.1 | ||||
NM_001314045.1 | Intron | NP_001300974.1 | ||||
NM_001314046.1 | Intron | NP_001300975.1 | ||||
NM_001314047.1 | Intron | NP_001300976.1 | ||||
NM_001314048.1 | Intron | NP_001300977.1 | ||||
NM_033439.3 | Intron | NP_254274.1 | ||||
XM_011518061.2 | Intron | XP_011516363.1 | ||||
XM_017015285.1 | Intron | XP_016870774.1 |