Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATAATTTAGCCCCAATAAATACA[C/T]TGTCCACATAAAGACCAGTTACTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605513 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA2026 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIAA2026 - KIAA2026 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017969.2 | 8238 | Intron | NP_001017969.2 | |||
XM_005251385.2 | 8238 | Intron | XP_005251442.1 | |||
XM_006716730.3 | 8238 | Intron | XP_006716793.1 | |||
XM_011517759.2 | 8238 | UTR 3 | XP_011516061.1 | |||
XM_011517760.2 | 8238 | Intron | XP_011516062.1 | |||
XM_011517761.2 | 8238 | Intron | XP_011516063.1 | |||
XM_017014331.1 | 8238 | Intron | XP_016869820.1 | |||
XM_017014332.1 | 8238 | Intron | XP_016869821.1 | |||
XM_017014333.1 | 8238 | Intron | XP_016869822.1 | |||
XM_017014334.1 | 8238 | Intron | XP_016869823.1 | |||
XM_017014335.1 | 8238 | Intron | XP_016869824.1 | |||
XM_017014336.1 | 8238 | Intron | XP_016869825.1 |
MLANA - melan-A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005511.1 | 8238 | Intron | NP_005502.1 |