Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTGAATGTTGTTACCTTTTCC[C/T]TTTTATAGCTTTTCTTTGCAGGTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLGRKT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLGRKT - plasminogen receptor with a C-terminal lysine | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018465.3 | Intron | NP_060935.2 | ||||
XM_005251510.4 | Intron | XP_005251567.1 | ||||
XM_005251512.4 | Intron | XP_005251569.1 | ||||
XM_011517960.2 | Intron | XP_011516262.1 |