Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATCATTTTGTGCTTTCCTTTTT[C/T]TTGGCTTTATTATCTATTTATTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601337 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RFX3 - regulatory factor X3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282116.1 | 4447 | UTR 3 | NP_001269045.1 | |||
NM_001282117.1 | 4447 | Intron | NP_001269046.1 | |||
NM_002919.3 | 4447 | Intron | NP_002910.1 | |||
NM_134428.2 | 4447 | UTR 3 | NP_602304.1 | |||
XM_006716840.1 | 4447 | UTR 3 | XP_006716903.1 | |||
XM_006716843.1 | 4447 | UTR 3 | XP_006716906.1 | |||
XM_006716844.1 | 4447 | UTR 3 | XP_006716907.1 | |||
XM_006716845.2 | 4447 | UTR 3 | XP_006716908.1 | |||
XM_006716847.3 | 4447 | UTR 3 | XP_006716910.1 | |||
XM_011517998.1 | 4447 | UTR 3 | XP_011516300.1 | |||
XM_011517999.1 | 4447 | UTR 3 | XP_011516301.1 | |||
XM_011518000.1 | 4447 | UTR 3 | XP_011516302.1 | |||
XM_011518001.1 | 4447 | UTR 3 | XP_011516303.1 | |||
XM_017015000.1 | 4447 | UTR 3 | XP_016870489.1 | |||
XM_017015001.1 | 4447 | UTR 3 | XP_016870490.1 | |||
XM_017015002.1 | 4447 | UTR 3 | XP_016870491.1 |
Set Membership: |
HapMap |