Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602424 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMRT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DMRT1 - doublesex and mab-3 related transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021951.2 | Intron | NP_068770.2 | ||||
XM_006716732.1 | Intron | XP_006716795.1 | ||||
XM_011517770.1 | Intron | XP_011516072.1 | ||||
XM_011517771.1 | Intron | XP_011516073.1 | ||||
XM_011517772.1 | Intron | XP_011516074.1 | ||||
XM_011517773.2 | Intron | XP_011516075.1 | ||||
XM_017014374.1 | Intron | XP_016869863.1 | ||||
XM_017014375.1 | Intron | XP_016869864.1 |
Set Membership: |
HapMap |