Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTACGTTCTGTAAGATTTGTTTA[A/G]CGTTTTAGGCACTTTACGCATACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600062 | ||||||||||||||||||||
Literature Links: |
RAD23B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD23B - RAD23 homolog B, nucleotide excision repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244713.1 | Intron | NP_001231642.1 | ||||
NM_001244724.1 | Intron | NP_001231653.1 | ||||
NM_002874.4 | Intron | NP_002865.1 |