Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTAGTATGAAAGTACTAACTTA[C/T]TGGTATATGGGTTTTGAAATAATAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600062 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD23B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAD23B - RAD23 homolog B, nucleotide excision repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244713.1 | Intron | NP_001231642.1 | ||||
NM_001244724.1 | Intron | NP_001231653.1 | ||||
NM_002874.4 | Intron | NP_002865.1 |
Set Membership: |
HapMap |