Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAGCATAGTAGGAAGAAGGAAT[C/T]ATCAGGGCAAGGCACACTGCAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616694 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0368 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0368 - KIAA0368 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080398.1 | 6242 | Intron | NP_001073867.1 | |||
XM_005251850.3 | 6242 | Intron | XP_005251907.1 | |||
XM_005251851.4 | 6242 | Intron | XP_005251908.1 | |||
XM_005251852.4 | 6242 | Intron | XP_005251909.1 | |||
XM_005251853.3 | 6242 | Intron | XP_005251910.1 | |||
XM_005251854.3 | 6242 | Intron | XP_005251911.1 | |||
XM_011518422.2 | 6242 | UTR 3 | XP_011516724.1 | |||
XM_017014557.1 | 6242 | Intron | XP_016870046.1 |