Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGACACCATCCAGAGACTAAA[A/C]CTTTGCTCAACAGAAATTTTAGAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf147 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C9orf147 - chromosome 9 open reading frame 147 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSDL2 - hydroxysteroid dehydrogenase like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA1958 - KIAA1958 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |