Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGACAGTGAGGTTCTTGGTAGGT[C/T]TTTGATTTGACCTTCCTCTAAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300024 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORA11C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SNORA11C - small nucleolar RNA, H/ACA box 11C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF157 - zinc finger protein 157 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003446.3 | Intron | NP_003437.2 |