Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTATGCTCTGAGAGTTTGGAATA[C/G]AACAGTCAACTTAGCCTGGATATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605261 | ||||||||||||||||||||
Literature Links: |
NOX4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOX4 - NADPH oxidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143836.2 | Intron | NP_001137308.1 | ||||
NM_001143837.1 | Intron | NP_001137309.1 | ||||
NM_001291926.1 | Intron | NP_001278855.1 | ||||
NM_001291927.1 | Intron | NP_001278856.1 | ||||
NM_001291929.1 | Intron | NP_001278858.1 | ||||
NM_001300995.1 | Intron | NP_001287924.1 | ||||
NM_016931.4 | Intron | NP_058627.1 | ||||
XM_006718849.3 | Intron | XP_006718912.1 | ||||
XM_011542857.2 | Intron | XP_011541159.1 | ||||
XM_017017841.1 | Intron | XP_016873330.1 | ||||
XM_017017842.1 | Intron | XP_016873331.1 | ||||
XM_017017843.1 | Intron | XP_016873332.1 | ||||
XM_017017844.1 | Intron | XP_016873333.1 | ||||
XM_017017845.1 | Intron | XP_016873334.1 |