Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600157 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1B1 - adaptor related protein complex 1 beta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127.3 | Intron | NP_001118.3 | ||||
NM_001166019.1 | Intron | NP_001159491.1 | ||||
NM_145730.2 | Intron | NP_663782.2 |
MIR3653 - microRNA 3653 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD125 - small nucleolar RNA, C/D box 125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |