Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611446 | ||||||||||||||||||||
Literature Links: |
DUSP18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP18 - dual specificity phosphatase 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304794.1 | Intron | NP_001291723.1 | ||||
NM_001304795.1 | Intron | NP_001291724.1 | ||||
NM_001304796.1 | Intron | NP_001291725.1 | ||||
NM_152511.4 | Intron | NP_689724.3 | ||||
XM_005261368.4 | Intron | XP_005261425.1 | ||||
XM_006724148.3 | Intron | XP_006724211.1 | ||||
XM_011529920.2 | Intron | XP_011528222.1 | ||||
XM_011529921.2 | Intron | XP_011528223.1 | ||||
XM_017028627.1 | Intron | XP_016884116.1 | ||||
XM_017028628.1 | Intron | XP_016884117.1 |
SLC35E4 - solute carrier family 35 member E4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001479.3 | Intron | NP_001001479.1 | ||||
NM_001318370.1 | Intron | NP_001305299.1 | ||||
NM_001318371.1 | Intron | NP_001305300.1 | ||||
XM_017028794.1 | Intron | XP_016884283.1 | ||||
XM_017028795.1 | Intron | XP_016884284.1 |