Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCACTCCACAGGAACAGCTGCA[C/G]CCACCCTGGCTGCACTCCTCGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605969 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RFPL2 - ret finger protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098527.2 | 719 | UTR 5 | NP_001091997.2 | |||
NM_001159545.1 | 719 | Intron | NP_001153017.1 | |||
NM_001159546.1 | 719 | Intron | NP_001153018.1 | |||
NM_006605.3 | 719 | Intron | NP_006596.2 | |||
XM_011529826.1 | 719 | UTR 5 | XP_011528128.1 | |||
XM_011529827.2 | 719 | Intron | XP_011528129.1 | |||
XM_011529828.1 | 719 | UTR 5 | XP_011528130.1 | |||
XM_011529830.2 | 719 | Intron | XP_011528132.1 | |||
XM_011529832.1 | 719 | UTR 5 | XP_011528134.1 | |||
XM_017028535.1 | 719 | Intron | XP_016884024.1 | |||
XM_017028536.1 | 719 | Intron | XP_016884025.1 |