Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604518 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRAP2 - GRB2-related adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291824.1 | Intron | NP_001278753.1 | ||||
NM_001291825.1 | Intron | NP_001278754.1 | ||||
NM_001291826.1 | Intron | NP_001278755.1 | ||||
NM_001291828.1 | Intron | NP_001278757.1 | ||||
NM_004810.3 | Intron | NP_004801.1 | ||||
XM_006724376.3 | Intron | XP_006724439.1 |